Tisk…

Tridecanoic acid 99%

Dodavatel: Apollo Scientific
Varování

OR54705-100G OR54705-25G OR54705-5G
APOSOR54705-100GEA 7770 CZK
APOSOR54705-100G APOSOR54705-25G APOSOR54705-5G
Tridecanoic acid 99%
Tridecanoic acid
Vzorec: C₁₃H₂₆O₂
Storage Temperature: Freezer
MDL Number: MFCD00002741
CAS číslo: 638-53-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR