Tisk…
Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl) 95%
Dodavatel: Apollo Scientific
Varování
Synonyms: H-Gly-OBzl·HCl, H-DL-Gly-OBzl hydrochloride, Benzyl glycinate hydrochloride, H-DL-Gly-OBzl.HCl
OR54703-100G OR54703-25G OR54703-5G OR54703-500G
APOSOR54703-100GEA 2830 CZK
APOSOR54703-100G APOSOR54703-25G APOSOR54703-5G APOSOR54703-500G
Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl) 95%
Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl)
Vzorec: C₉H₁₂ClNO₂
MW: 201.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00035442
CAS číslo: 2462-31-9