Tisk…

Propiolaldehyde diethyl acetal 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: Propargylaldehyde diethyl acetal, 3,3-Diethoxypropyne, Propioaldehyde diethyl acetal, Propynal diethyl acetal, 3,3-Diethoxy-1-propyne, 3,3-Diethoxyprop-1-yne

OR54615-25G OR54615-5G OR54615-100G
APOSOR54615-25GEA 1240 CZK
APOSOR54615-25G APOSOR54615-5G APOSOR54615-100G
Propiolaldehyde diethyl acetal 95%
Propiolaldehyde diethyl acetal
Vzorec: C₇H₁₂O₂
MW: 128.17
Bod varu: 138…139.5 °C
Hustota: 0.894 g/cm³ (25 °C)
Bod vzplanutí: 31 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009237
CAS číslo: 10160-87-9
UN: 1993
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR