Tisk…

8-Bromo-5-nitroisoquinoline 98%

Dodavatel: Apollo Scientific
Varování

OR54593-1G OR54593-250MG OR54593-5G
APOSOR54593-1GEA 2440 CZK
APOSOR54593-1G APOSOR54593-250MG APOSOR54593-5G
8-Bromo-5-nitroisoquinoline 98%
8-Bromo-5-nitroisoquinoline
Vzorec: C₉H₅BrN₂O₂
MW: 253.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08282783
CAS číslo: 252861-41-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR