Tisk…

Methyl 3,5-Dihydroxy-4-methoxybenzoate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3,5-Dihydroxy-4-methoxybenzoic acid methyl ester, Methyl 3,5-dihydroxy-4-methoxybenzoate

OR54581-1G OR54581-5G
APOSOR54581-1GEA 2860 CZK
APOSOR54581-1G APOSOR54581-5G
Methyl 3,5-Dihydroxy-4-methoxybenzoate 98%
Methyl 3,5-Dihydroxy-4-methoxybenzoate
Vzorec: C₉H₁₀O₅
MW: 198.17 g/mol
Bod tání: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00017195
CAS číslo: 24093-81-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR