Tisk…

Fmoc-Pro-OH 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-proline, N-Fmoc-L-proline

OR54567-100G OR54567-1KG OR54567-500G
APOSOR54567-100GEA 2690 CZK
APOSOR54567-100G APOSOR54567-1KG APOSOR54567-500G
Fmoc-Pro-OH 98%
Fmoc-Pro-OH
Vzorec: C₂₀H₁₉NO₄
Bod tání: 117…118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037122
CAS číslo: 71989-31-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR