Tisk…

1-Boc-4-(6-nitro-3-pyridyl)piperazine 97%

Dodavatel: Apollo Scientific

Synonyms: tert-Butyl 4-(6-nitropyridin-3-yl)piperazine-1-carboxylate, 4-(6-Nitropyridin-3-yl)piperazine, N1-BOC protected

OR54564-10G OR54564-5G
APOSOR54564-10GEA 4530 CZK
APOSOR54564-10G APOSOR54564-5G
1-Boc-4-(6-nitro-3-pyridyl)piperazine 97%
1-Boc-4-(6-nitro-3-pyridyl)piperazine

Intermediate in Ribociclib synthesis.

Vzorec: C₁₄H₂₀N₄O₄
MW: 308,33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11849291
CAS číslo: 571189-16-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR