Phthalimide potassium salt 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
N-Potassium phthalimide, Potassium 1,3-dioxoisoindolin-2-ide, Potassium phthalimide
OR54559-500G
OR54559-100G
APOSOR54559-500GEA
1660
CZK
APOSOR54559-500G
APOSOR54559-100G
Phthalimide potassium salt 97%
Phthalimide potassium salt
Vzorec:
C₈H₄KNO₂ MW: 185.22 Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005887 CAS číslo: 1074-82-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...