N,N-Dimethyl-1,3-diaminopropan 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3-Dimethylaminopropylamine, N,N-Dimethyltrimethylenediamine, N,N-Dimethyl-1,3-propanediamine
OR54483-100G
OR54483-1KG
OR54483-500G
APOSOR54483-100GEA
3440
CZK
APOSOR54483-100G
APOSOR54483-1KG
APOSOR54483-500G
N,N-Dimethyl-1,3-diaminopropan 95%
N,N-Dimethyl-1,3-diaminopropan
Vzorec:
C₅H₁₄N₂ MW: 102,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00008216 CAS číslo: 109-55-7 UN: 2734 ADR: 8+3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...