Octadecanal 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Octadecyl Aldehyde, Octadecanaldehyde, Stearyl Aldehyde, Stearaldehyde
OR54467-1G
OR54467-5G
APOSOR54467-1GEA
1580
CZK
APOSOR54467-1G
APOSOR54467-5G
Octadecanal 95%
Octadecanal
Vzorec:
C₁₈H₃₆O MW: 268,49 g/mol Bod varu: 215 °C/20mmHg Bod tání: 46 °C Storage Temperature: Freezer |
MDL Number:
MFCD03790007 CAS číslo: 638-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...