Ethyl all-cis-5,8,11,14,17-eicosapentaenoate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
EPA Ethyl Ester, all cis-5,8,11,14,17-Eicosapentaenoic Acid Ethyl Ester, Ethyl all cis-5,8,11,14,17-Icosapentaenoate, Ethyl (5Z,8Z,11Z,14Z,17Z)-icosa-5,8,11,14,17-pentaenoate
OR54466-100MG
OR54466-50MG
APOSOR54466-100MGEA
9330
CZK
APOSOR54466-100MG
APOSOR54466-50MG
Ethyl all-cis-5,8,11,14,17-eicosapentaenoate
Ethyl all-cis-5,8,11,14,17-eicosapentaenoate
Vzorec:
C₂₂H₃₄O₂ MW: 330,51 g/mol Hustota: 0,91 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00673476 CAS číslo: 86227-47-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...