Tisk…

β-Alanine-tert-butylester hydrochloride 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: beta-Alanine tert-Butyl ester hydrochloride, β-Alanine t-butyl ester hydrochloride, H-β-Ala-OtBu·HCl, H-beta-Ala-OtBu·HCl, H-beta-Ala-OtBu.HCl, H-β-Ala-OtBu.HCl, tert-Butyl 3-aminopropanoate hydrochloride

OR54450-100G OR54450-500G OR54450-25G
APOSOR54450-100GEA 12470 CZK
APOSOR54450-100G APOSOR54450-500G APOSOR54450-25G
β-Alanine-tert-butylester hydrochloride 97%
beta-Alanine-tert-butylester hydrochloride
Vzorec: C₇H₁₆ClNO₂
MW: 181,66 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00058258
CAS číslo: 58620-93-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR