Tisk…

N-1H-Benzimidazol-2-ylguanidine

Dodavatel: Apollo Scientific

Synonyms: 2-Guanidinobenzimiazole

OR54411-1G OR54411-5G
APOSOR54411-1GEA 1990 CZK
APOSOR54411-1G APOSOR54411-5G
N-1H-Benzimidazol-2-ylguanidine
N-1H-Benzimidazol-2-ylguanidine
Vzorec: C₈H₉N₅
MW: 175,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005590
CAS číslo: 5418-95-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR