2-Iodopyrimidine 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR54388-1G
OR54388-5G
OR54388-25G
APOSOR54388-1GEA
1040
CZK
APOSOR54388-1G
APOSOR54388-5G
APOSOR54388-25G
2-Iodopyrimidine 95%
2-Iodopyrimidine
Vzorec:
C₄H₃IN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD09835340 CAS číslo: 31462-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...