Tisk…

4-(Pyrrolidin-3-yl)morpholine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-(3-Pyrrolidinyl)morpholine

OR54379-1G OR54379-5G
APOSOR54379-1GEA 5590 CZK
APOSOR54379-1G APOSOR54379-5G
4-(Pyrrolidin-3-yl)morpholine
4-(Pyrrolidin-3-yl)morpholine
Vzorec: C₈H₁₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD06740352
CAS číslo: 53617-37-1
UN: 2735
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR