(3-Chloropyrazin-2-yl)methanamine dihydrochloride 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(Aminomethyl)-3-chloropyrazine dihydrochloride
OR54357-1G
OR54357-500MG
APOSOR54357-1GEA
2120
CZK
APOSOR54357-1G
APOSOR54357-500MG
(3-Chloropyrazin-2-yl)methanamine dihydrochloride 95%
(3-Chloropyrazin-2-yl)methanamine dihydrochloride
Vzorec:
C₅H₈Cl₃N₃ MW: 216,5 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07644629 CAS číslo: 867165-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...