Tisk…
1,5-Naphthalenedisulphonic acid tetrahydrate 98%
Dodavatel: Apollo Scientific
Nebezpečí
Synonyms: Naphthalene-1,5-disulphonic acid tetrahydrate
OR54356-100G OR54356-25G OR54356-500G
APOSOR54356-100GEA 1740 CZK
APOSOR54356-100G APOSOR54356-25G APOSOR54356-500G
1,5-Naphthalenedisulphonic acid tetrahydrate 98%
1,5-Naphthalenedisulphonic acid tetrahydrate
Vzorec: C₁₀H₁₆O₁₀S₂
MW: 360.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149265
CAS číslo: 211366-30-2
EINECS: 201-317-9
UN: 2585
ADR: 8 III