Tisk…

(3R,4R)-1-Benzyl-N,4-dimethylpiperidin-3-amine dihydrochloride 95+%

Dodavatel: Apollo Scientific

Synonyms: cis-1-Benzyl-N3,4-dimethylpiperidin-3-amine dihydrochloride, cis-1-Benzyl-4-methyl-3-(methylamino)piperidine dihydrochloride

OR54340-100G OR54340-25G
APOSOR54340-100GEA 11180 CZK
APOSOR54340-100G APOSOR54340-25G
(3R,4R)-1-Benzyl-N,4-dimethylpiperidin-3-amine dihydrochloride 95+%
(3R,4R)-1-Benzyl-N,4-dimethylpiperidin-3-amine dihydrochloride
Vzorec: C₁₄H₂₄Cl₂N₂
MW: 291,26 g/mol
Bod tání: 249…251 °C
Storage Temperature: Ambient
MDL Number: MFCD11044501
CAS číslo: 477600-68-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR