Tisk…

Methyl-4-(methylamino)-3-nitrobenzenecarboxylate

Dodavatel: Apollo Scientific

Synonyms: Methyl 4-(methylamino)-3-nitrobenzoate

OR54327-5G
APOSOR54327-5GEA 2900 CZK
APOSOR54327-5G
Methyl-4-(methylamino)-3-nitrobenzenecarboxylate
Methyl-4-(methylamino)-3-nitrobenzenecarboxylate
Vzorec: C₉H₁₀N₂O₄
MW: 210,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09862450
CAS číslo: 36242-50-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR