4-Bromo-2-iodoaniline
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR5425-25G
OR5425-5G
OR5425-1G
OR5425-100G
APOSOR5425-25GEA
1240
CZK
APOSOR5425-25G
APOSOR5425-5G
APOSOR5425-1G
APOSOR5425-100G
4-Bromo-2-iodoaniline
4-Bromo-2-iodoaniline
Vzorec:
C₆H₅BrIN MW: 297.92 Bod tání: 69…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093764 CAS číslo: 66416-72-6 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...