Tisk…

4-Bromo-3-chloroanisole 98%

Dodavatel: Apollo Scientific
Varování

OR5424-1G OR5424-5G OR5424-25G OR5424-100G
APOSOR5424-1GEA 417 CZK
APOSOR5424-1G APOSOR5424-5G APOSOR5424-25G APOSOR5424-100G
4-Bromo-3-chloroanisole 98%
4-Bromo-3-chloroanisole
Vzorec: C₇H₆BrClO
MW: 221,48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070742
CAS číslo: 50638-46-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR