Tisk…

Tetrabutylammonium tetrahydroborate

Dodavatel: Apollo Scientific

Synonyms: Tetra-n-butylammonium borohydride, Tetrabutylammonium borohydride, Tetrabutylammonium tetrahydridoborate

OR5417-50G OR5417-100G
APOSOR5417-50GEA 6450 CZK
APOSOR5417-50G APOSOR5417-100G
Tetrabutylammonium tetrahydroborate
Tetrabutylammonium tetrahydroborate
Vzorec: C₁₆H₄₀BN
MW: 257,31 g/mol
Bod tání: 126…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00012035
CAS číslo: 33725-74-5
UN: 3131
ADR: 4.3+8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR