Tisk…

Sulphur trioxide trimethylamine complex 95%

Dodavatel: Apollo Scientific
Nebezpečí

OR5415-100G OR5415-25G
APOSOR5415-100GEA 2320 CZK
APOSOR5415-100G APOSOR5415-25G
Sulphur trioxide trimethylamine complex 95%
Sulphur trioxide trimethylamine complex
Vzorec: C₃H₉NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00012421
CAS číslo: 3162-58-1
EINECS: 221-614-7
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR