Tisk…

3-Biphenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: [1,1'-Biphenyl]-3-ylboronic acid

OR5411-1G OR5411-25G OR5411-5G
APOSOR5411-1GEA 622 CZK
APOSOR5411-1G APOSOR5411-25G APOSOR5411-5G
3-Biphenylboronic acid
3-Biphenylboronic acid
Vzorec: C₁₂H₁₁BO₂
MW: 198,03 g/mol
Bod tání: 206…211 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01318102
CAS číslo: 5122-95-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR