3-[4-(tert-Butoxycarbonyl)piperazin-1-yl]propanoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR5406-1G
OR5406-250MG
OR5406-5G
APOSOR5406-1GEA
3740
CZK
APOSOR5406-1G
APOSOR5406-250MG
APOSOR5406-5G
3-[4-(tert-Butoxycarbonyl)piperazin-1-yl]propanoic acid
3-[4-(tert-Butoxycarbonyl)piperazin-1-yl]propanoic acid
Vzorec:
C₁₂H₂₂N₂O₄ Bod tání: 144…145 °C Storage Temperature: Ambient |
MDL Number:
MFCD03410257 CAS číslo: 242459-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...