Tisk…

1-Benzyl-4-cyano-4-hydroxypiperidine hydrochloride 98%

Dodavatel: Apollo Scientific
Nebezpečí

OR5395-100G OR5395-25G OR5395-5G
APOSOR5395-100GEA 10580 CZK
APOSOR5395-100G APOSOR5395-25G APOSOR5395-5G
1-Benzyl-4-cyano-4-hydroxypiperidine hydrochloride 98%
1-Benzyl-4-cyano-4-hydroxypiperidine hydrochloride
Vzorec: C₁₃H₁₇ClN₂O
Bod tání: 176 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00012836
CAS číslo: 71617-20-4
EINECS: 275-713-5
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR