4-Nitrobenzyl chloroformate 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-Nitrobenzyl carbonochloridate
OR5386-100G
OR5386-25G
OR5386-5G
APOSOR5386-100GEA
7670
CZK
APOSOR5386-100G
APOSOR5386-25G
APOSOR5386-5G
4-Nitrobenzyl chloroformate 97%
4-Nitrobenzyl chloroformate
Vzorec:
C₈H₆ClNO₄ Bod varu: 230 °C/9,75mmHg Bod tání: 32…34 °C Bod vzplanutí: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007375 CAS číslo: 4457-32-3 EINECS: 224-708-6 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...