2,5-Dichloro-3-thiophenesulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR5385-1G
OR5385-5G
APOSOR5385-1GEA
2280
CZK
APOSOR5385-1G
APOSOR5385-5G
2,5-Dichloro-3-thiophenesulphonyl chloride
2,5-Dichloro-3-thiophenesulphonyl chloride
Vzorec:
C₄HCl₃O₂S₂ MW: 251,54 g/mol Bod varu: 256…257 °C Hustota: 1,697 g/cm³ (25 °C) Bod vzplanutí: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00051665 CAS číslo: 56946-83-9 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...