3-Bromobenzenesulphonyl chloride 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR5371-100G
OR5371-25G
APOSOR5371-100GEA
3740
CZK
APOSOR5371-100G
APOSOR5371-25G
3-Bromobenzenesulphonyl chloride 97%
3-Bromobenzenesulphonyl chloride
Vzorec:
C₆H₄BrClO₂S MW: 255,52 g/mol Bod varu: 90…91 °C/0,5mmHg Bod tání: 30…33 °C Hustota: 1,773 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00052313 CAS číslo: 2905-24-0 UN: 3265 ADR: 8,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...