4-Methoxybenzophenone 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
p-Methoxybenzophenone, (4-Methoxyphenyl)(phenyl)methanone
OR5356-100G
OR5356-25G
OR5356-500G
APOSOR5356-100GEA
787
CZK
APOSOR5356-100G
APOSOR5356-25G
APOSOR5356-500G
4-Methoxybenzophenone 98%
4-Methoxybenzophenone
Vzorec:
C₁₄H₁₂O₂ MW: 212,25 g/mol Bod varu: 354…356 °C Bod tání: 60…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008403 CAS číslo: 611-94-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...