Tisk…

o-Anisic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Methoxybenzoic acid

OR5344-10KG OR5344-1KG OR5344-5KG
APOSOR5344-10KGEA 21490 CZK
APOSOR5344-10KG APOSOR5344-1KG APOSOR5344-5KG
o-Anisic acid 98%
o-Anisic acid
Vzorec: C₈H₈O₃
MW: 152,15 g/mol
Bod varu: 200 °C
Bod tání: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002431
CAS číslo: 579-75-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR