4-(2-Nitroethyl)phenylboronic acid 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR5321-1G
OR5321-250MG
APOSOR5321-1GEA
21490
CZK
APOSOR5321-1G
APOSOR5321-250MG
4-(2-Nitroethyl)phenylboronic acid 96%
4-(2-Nitroethyl)phenylboronic acid
Vzorec:
C₈H₁₀BNO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD09258738 CAS číslo: 957034-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...