Phenyl acetate 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Acetic acid phenyl ester
OR5316-100G
OR5316-250G
OR5316-1KG
APOSOR5316-100GEA
622
CZK
APOSOR5316-100G
APOSOR5316-250G
APOSOR5316-1KG
Phenyl acetate 97%
Phenyl acetate
Vzorec:
C₈H₈O₂ MW: 136.15 Bod varu: 196 °C Hustota: 1.075 g/cm³ (25 °C) Bod vzplanutí: 76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008699 CAS číslo: 122-79-2 EINECS: 204-575-0 Merck Index: 12,07421 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...