Tisk…
Pyruvaldehyde (methylglyoxal) 35% in aqueous solution
Dodavatel: Apollo Scientific
Nebezpečí
Synonyms: Pyruvicaldehyde, Pyruvic aldehyde
OR53061-100G OR53061-1KG OR53061-500G
APOSOR53061-100GEA 1040 CZK
APOSOR53061-100G APOSOR53061-1KG APOSOR53061-500G
Pyruvaldehyde (methylglyoxal) 35% in aqueous solution
Pyruvaldehyde (methylglyoxal)
Vzorec: C₃H₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00006960
CAS číslo: 78-98-8