Ethyl cyclopropanecarboxylate 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR5300-25G
OR5300-100G
OR5300-500G
APOSOR5300-25GEA
622
CZK
APOSOR5300-25G
APOSOR5300-100G
APOSOR5300-500G
Ethyl cyclopropanecarboxylate 98%
Ethyl cyclopropanecarboxylate
Vzorec:
C₆H₁₀O₂ MW: 114,15 g/mol Bod varu: 133…134 °C Hustota: 0,96 g/cm³ (25 °C) Bod vzplanutí: 18 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001282 CAS číslo: 4606-07-9 EINECS: 225-010-4 UN: 3272 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...