Tisk…

4,6-Dimethyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine

Dodavatel: Apollo Scientific
Varování

OR52981-10G OR52981-1G OR52981-5G
APOSOR52981-10GEA 8270 CZK
APOSOR52981-10G APOSOR52981-1G APOSOR52981-5G
4,6-Dimethyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine
4,6-Dimethyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine
Vzorec: C₈H₁₀N₄
MW: 162,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00124667
CAS číslo: 41601-44-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR