Tisk…

2-Acetoxyisobutyroyl bromide 96%

Dodavatel: Apollo Scientific

OR52938-100G OR52938-25G OR52938-5G
APOSOR52938-100GEA 10060 CZK
APOSOR52938-100G APOSOR52938-25G APOSOR52938-5G
2-Acetoxyisobutyroyl bromide 96%
2-Acetoxyisobutyroyl bromide
Vzorec: C₆H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD00040923
CAS číslo: 40635-67-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR