Methyl-1-hydroxy-2-naphthoate
Dodavatel: Apollo Scientific
Synonyms:
1-Hydroxy-2-naphthoic acid methyl ester, 1-Hydroxy-β-naphthoic acid methyl ester, Methyl 1-hydroxy-2-naphthoate
OR52903-1G
OR52903-5G
APOSOR52903-1GEA
4770
CZK
APOSOR52903-1G
APOSOR52903-5G
Methyl-1-hydroxy-2-naphthoate
Methyl-1-hydroxy-2-naphthoate
Vzorec:
C₁₂H₁₀O₃ MW: 202,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00043823 CAS číslo: 948-03-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...