Methyl-3-bromo-4-methoxybenzoate
Dodavatel: Apollo Scientific
Synonyms:
Methyl 3-bromo-4-methoxybenzoate, 3-Bromo-4-methoxybenzoic acid methyl ester
OR52901-25G
APOSOR52901-25GEA
5560
CZK
APOSOR52901-25G
Methyl-3-bromo-4-methoxybenzoate
Methyl-3-bromo-4-methoxybenzoate
Vzorec:
C₉H₉BrO₃ MW: 245,07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00210463 CAS číslo: 35450-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...