Tisk…

Methyl-4-phenylbenzoate

Dodavatel: Apollo Scientific

Synonyms: p-Phenylbenzoic acid-OMe, Methyl biphenyl-4-carboxylate, Methyl [1,1'-biphenyl]-4-carboxylate

OR52898-100G
APOSOR52898-100GEA 10390 CZK
APOSOR52898-100G
Methyl-4-phenylbenzoate
Methyl 4-phenylbenzoate
Vzorec: C₁₄H₁₂O₂
MW: 212,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017200
CAS číslo: 720-75-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR