Tisk…

(±)-α-Bromo-γ-butyrolactone 95+%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Bromo-γ-butyrolactone, (±)-2-Bromobutan-4-olide, 2-Bromo-4-butanolide, (±)-2-Bromo-4-hydroxybutanoic acid lactone, (±)-2-Bromo-4-butanolide

OR5280-100G OR5280-500G
APOSOR5280-100GEA 1990 CZK
APOSOR5280-100G APOSOR5280-500G
(±)-α-Bromo-γ-butyrolactone 95+%
(±)-α-Bromo-γ-butyrolactone
Vzorec: C₄H₅BrO₂
MW: 164.99
Bod varu: 138 °C/6mmHg
Hustota: 1.786 g/cm³ (25 °C)
Bod vzplanutí: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005387
CAS číslo: 5061-21-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR