4-Piperidin-1-ylpyridin-2-amine 95%
Dodavatel: Apollo Scientific
OR52771-1G
OR52771-250MG
APOSOR52771-1GEA
25630
CZK
APOSOR52771-1G
APOSOR52771-250MG
4-Piperidin-1-ylpyridin-2-amine 95%
4-Piperidin-1-ylpyridin-2-amine
Vzorec:
C₁₀H₁₅N₃ Storage Temperature: Ambient |
MDL Number:
MFCD01646275 CAS číslo: 52311-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...