Tisk…

1-(4-Aminophenyl)piperazine 96%

Dodavatel: Apollo Scientific

Synonyms: 4-Piperazin-1-ylaniline

OR52766-1G OR52766-5G
APOSOR52766-1GEA 1740 CZK
APOSOR52766-1G APOSOR52766-5G
1-(4-Aminophenyl)piperazine 96%
1-(4-Aminophenyl)piperazine
Vzorec: C₁₀H₁₅N₃
Storage Temperature: Refrigerator
MDL Number: MFCD01365906
CAS číslo: 67455-41-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR