6-Bromo-4-chloroquinazoline
Dodavatel: Apollo Scientific
|
Varování
|
OR52695-25G
OR52695-5G
APOSOR52695-25GEA
3610
CZK
APOSOR52695-25G
APOSOR52695-5G
6-Bromo-4-chloroquinazoline
6-Bromo-4-chloroquinazoline
Vzorec:
C₈H₄BrClN₂ MW: 243,4878 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01862191 CAS číslo: 38267-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...