Tisk…

(3-Chloro-4-methoxyphenyl)methanamine

Dodavatel: Apollo Scientific
Nebezpečí

OR52670-1G OR52670-5G
APOSOR52670-1GEA 2490 CZK
APOSOR52670-1G APOSOR52670-5G
(3-Chloro-4-methoxyphenyl)methanamine
(3-Chloro-4-methoxyphenyl)methanamine
Vzorec: C₈H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD00995395
CAS číslo: 115514-77-7
UN: 1760
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR