Tisk…

(R)-3-Iodo-tetrahydrofuran

Dodavatel: Apollo Scientific
Varování

OR52586-1G OR52586-250MG OR52586-5G
APOSOR52586-1GEA 3440 CZK
APOSOR52586-1G APOSOR52586-250MG APOSOR52586-5G
(R)-3-Iodo-tetrahydrofuran
(R)-3-Iodo-tetrahydrofuran
Vzorec: C₄H₇IO
Storage Temperature: Ambient
MDL Number: MFCD17015957
CAS číslo: 873063-62-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR