3-Iodotetrahydrofuran
Dodavatel: Apollo Scientific
|
Varování
|
OR52585-1G
OR52585-5G
APOSOR52585-1GEA
1050
CZK
APOSOR52585-1G
APOSOR52585-5G
3-Iodotetrahydrofuran
3-Iodotetrahydrofuran
Vzorec:
C₄H₇IO Bod varu: 110 °C/30mmHg Storage Temperature: Ambient |
MDL Number:
MFCD09878873 CAS číslo: 121138-01-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...