Tisk…

2-Cyanoethyl acrylate

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Acrylic Acid 2-Cyanoethyl Ester, 3-Acryloyloxypropionitrile

OR52542-25G OR52542-5G
APOSOR52542-25GEA 16170 CZK
APOSOR52542-25G APOSOR52542-5G
2-Cyanoethyl acrylate
2-Cyanoethyl acrylate
Vzorec: C₆H₇NO₂
MW: 125,13 g/mol
Bod varu: 180 °C/2mmHg
Hustota: 1,07 g/cm³ (25 °C)
Bod vzplanutí: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD00013823
CAS číslo: 106-71-8
EINECS: 203-426-7
UN: 3276
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR