H-Glu(OtBu)-OtBu·HCl 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
H-Glu(OtBu)-OtBu·HCl, L-Glutamic acid di-tert-butyl ester hydrochloride, H-Glu(OtBu)-OtBu.HCl
OR52525-25G
OR52525-5G
OR52525-100G
APOSOR52525-25GEA
2780
CZK
APOSOR52525-25G
APOSOR52525-5G
APOSOR52525-100G
H-Glu(OtBu)-OtBu·HCl 98%
H-Glu(OtBu)-OtBu·HCl
Vzorec:
C₁₃H₂₆ClNO₄ MW: 295,804 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00058003 CAS číslo: 32677-01-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...