Tisk…

Methyl-4-amino-1-methyl-1H-pyrrole-2-carboxylate hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 4-amino-1-methyl-1H-pyrrole-2-carboxylate hydrochloride, 4-Amino-1-methyl-1H-pyrrole-2-carboxylic acid-methyl ester·HCl

OR52508-1G OR52508-5G
APOSOR52508-1GEA 2530 CZK
APOSOR52508-1G APOSOR52508-5G
Methyl-4-amino-1-methyl-1H-pyrrole-2-carboxylate hydrochloride
Methyl-4-amino-1-methyl-1H-pyrrole-2-carboxylate hydrochloride
Vzorec: C₇H₁₁ClN₂O₂
MW: 190.627 g/mol
Bod tání: 236 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD01318761
CAS číslo: 180258-45-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR